Learn More
Leybold Vacuum Products Inc Leybonol LVO 420 Synthetic Oil, 1 L
Catalog No. NC1175480
Encompass_Preferred
Synthetic oil (perfluoropolyether PFPE, free of Additivs). Chemically inert. Highest thermal stability. Pumping axidants like oxygen, ozone or nitrous oxides as well as reactive substances like halogens, hydrogen halides and conditionally Lewis acids.
Catalog No. NC1175480 Supplier Leybold Vacuum Products Inc Supplier No. L42001
Synthetic oil (perfluoropolyether PFPE, free of Additivs). Chemically inert. Highest thermal stability. Pumping axidants like oxygen, ozone or nitrous oxides as well as reactive substances like halogens, hydrogen halides and conditionally Lewis acids.