Learn More
Accuflow Systems Saint-Gobain Tubing Connector, PVDF, Reducer, 1/8 in to 1/4 in
Catalog No. NC3363119
Encompass_Preferred
The Saint-Gobain Pure-Fit SIB tubing connector is a PVDF smooth inner-bore hose-barb reducer designed to provide a seamless transition between tubing sizes while minimizing flow disturbance. It is intended for reliable tubing-to-fitting connections and is packaged for bulk installation use.
  • Smooth inner-bore hose-barb design for seamless transitions between tubing and fittings.
  • Reducer configuration for 1/8 in to 1/4 in tubing connections.
  • Made from PVDF (polyvinylidene fluoride) for chemical resistance and purity.
  • Packaged 25 pieces per bag for bulk assembly.
  • Engineered for tubing-to-fitting fluid transfer applications where a smooth internal bore is required.
Catalog No. NC3363119 Supplier Accuflow Systems Supplier No. PFR125X250PVDF
The Saint-Gobain Pure-Fit SIB tubing connector is a PVDF smooth inner-bore hose-barb reducer designed to provide a seamless transition between tubing sizes while minimizing flow disturbance. It is intended for reliable tubing-to-fitting connections and is packaged for bulk installation use.
  • Smooth inner-bore hose-barb design for seamless transitions between tubing and fittings.
  • Reducer configuration for 1/8 in to 1/4 in tubing connections.
  • Made from PVDF (polyvinylidene fluoride) for chemical resistance and purity.
  • Packaged 25 pieces per bag for bulk assembly.
  • Engineered for tubing-to-fitting fluid transfer applications where a smooth internal bore is required.