Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Connector, PVDF, Reducer, 3/8 to 1/2 in
Catalog No. NC3363120
Encompass_Preferred
The Pure-Fit SIB reducer is a smooth inner-bore PVDF tubing connector designed to provide leak-free, low-dead-volume transitions between 3/8 in and 1/2 in flexible tubing for biopharmaceutical and laboratory fluid systems. It is manufactured in a certified clean room and intended for sterile fluid-transfer applications where chemical compatibility and fluid integrity are required.
  • Smooth inner bore reduces dead volume and entrapment.
  • PVDF construction provides chemical resistance and high purity.
  • Fits 3/8 in to 1/2 in tubing (9.5 x 12.7 mm).
  • Manufactured and packaged in a certified clean room.
  • Suitable for sterile fluid transfer and low-dead-volume connections.
Catalog No. NC3363120 Supplier Accuflow Systems Supplier No. PFR375X500PVDF
The Pure-Fit SIB reducer is a smooth inner-bore PVDF tubing connector designed to provide leak-free, low-dead-volume transitions between 3/8 in and 1/2 in flexible tubing for biopharmaceutical and laboratory fluid systems. It is manufactured in a certified clean room and intended for sterile fluid-transfer applications where chemical compatibility and fluid integrity are required.
  • Smooth inner bore reduces dead volume and entrapment.
  • PVDF construction provides chemical resistance and high purity.
  • Fits 3/8 in to 1/2 in tubing (9.5 x 12.7 mm).
  • Manufactured and packaged in a certified clean room.
  • Suitable for sterile fluid transfer and low-dead-volume connections.