Learn More
Analytical Sales & Services PFSA Ion Exchange Channel Separator Membrane
Catalog No. 5000805108
Encompass
PFSA Ion Exchange Channel Separator Membrane\nfor use with Flow Electrolysis Dual Stream (divided) Cell\n\n• Thickness: .007"\n• Color: clear\n\nQty/Pkg: Each
Catalog No. 50-008-05108 Supplier Analytical Sales & Services Supplier No. F1088201
PFSA Ion Exchange Channel Separator Membrane\nfor use with Flow Electrolysis Dual Stream (divided) Cell\n\n• Thickness: .007"\n• Color: clear\n\nQty/Pkg: Each