Learn More
Synthware™ PFTE Sleeve
GREENER_CHOICESDP
Catalog No. 31501917 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
29/42
34/45
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-501-917 34/45
31-501-913 14/20
31-501-914 19/22
31-501-915 24/40
31-501-916 29/42
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 31-501-917 Supplier Synthware™ Supplier No. T133445
Add to Cart
Add to Cart
Ribbed with gripping ring
Specifications
Material PTFE
Quantity 1 Ea.
For Use With (Application) Organic Synthesis
Type Ribbed PTFE Sleeve
Standard Taper Joint No. 34/45