Learn More
INTACT GENOMICS INC Pfu 2x Master Mix 200 reactions
Supplier:  INTACT GENOMICS INC 3326
Encompass
Pfu DNA Polymerase 2x master mix is a ready to use premix which contains Pfu DNA Polymerase, dNTPs, MgCl2 and stabilizers with optimized reaction buffer. It has been optimized for routine PCR applications. Pfu DNA Polymerase is a heat stable DNA polymerase which has 5´→ 3´ DNA polymerase and 3´→ 5´ exonuclease (proofreading) activities. Pfu DNA polymerase retains the high fidelity, sensitivity and processivity with an error rate six-fold lower than Taq DNA polymerase, and significantly lower than the error rates of most other proofreading enzymes or DNA polymerase mixtures. Pfu 2x Master Mix product is supplied with the unique Intact Genomics 5x Magic Enhancer that enables efficient amplification of GC rich templates up to 84%.Product SourceE. coli strain expressing a Pfu DNA Polymerase gene from Pyrococcus furiosus.Applications Routine PCR Primer extension Colony PCR Genotyping Efficient for amplifying high GC content template DNA with Magic Enhancer
Catalog No. 50-000-82906