Learn More
LAMDA BIOTECH INC Pfu 2X Master Mix
Catalog No. NC3726064
Encompass_Preferred
A pre-formulated PCR master mix containing Pfu DNA polymerase with proofreading capability, designed for high-fidelity DNA amplification applications. Includes dual-color tracking dyes for visual monitoring of electrophoresis.
Catalog No. NC3726064 Supplier LAMDA BIOTECH INC Supplier No. D152
A pre-formulated PCR master mix containing Pfu DNA polymerase with proofreading capability, designed for high-fidelity DNA amplification applications. Includes dual-color tracking dyes for visual monitoring of electrophoresis.