Learn More
Agilent Technologies PFU DNA LIGASE
Catalog No. 50125932
Encompass
Pfu DNA Ligase; Stratagene; From Pyrococcus furiosus; Catalyzes linkage of adjacent 5 in.-phosphate and 3 in.-hydroxy ends of ds DNA at 45deg. to 80deg.C; Highly thermostable; w/half-life of over 60 min at 95deg.C
Catalog No. 50-125-932 Supplier Agilent Technologies Supplier No. 600191
Pfu DNA Ligase; Stratagene; From Pyrococcus furiosus; Catalyzes linkage of adjacent 5 in.-phosphate and 3 in.-hydroxy ends of ds DNA at 45deg. to 80deg.C; Highly thermostable; w/half-life of over 60 min at 95deg.C