Learn More
Agilent Technologies PFU DNA POLYMERASE
Catalog No. 50125916
Encompass
Pfu DNA Polymerase 100 U Native version. The highest fidelity enzyme for PCR and primer-extension reactions PCR cloning and generating blunt-end amplification product Contains: Native Pfu DNA polymerase 10X Native Plus buffer
Catalog No. 50-125-916 Supplier Agilent Technologies Supplier No. 600135
Pfu DNA Polymerase 100 U Native version. The highest fidelity enzyme for PCR and primer-extension reactions PCR cloning and generating blunt-end amplification product Contains: Native Pfu DNA polymerase 10X Native Plus buffer