Learn More
Meridian Life Science Pfu DNA Polymerase Enzyme Only, 10ML
SDP
Catalog No. 5000856219
Encompass_Preferred
High-Fidelity Pfu is a thermostable high fidelity DNA polymerase enzyme, isolated from Pyrococcus furiosus, with separate 10x Pfu Reaction Buffer and MgCl2. The Pfu polymerase enzyme replicates DNA in the 5´-3´ direction in the presence of magnesium, but also possesses 3´-5´ exonuclease (proofreading) activity, making it ideal for NGS library amplification.
Catalog No. 50-008-56219 Supplier Meridian Life Science Supplier No. MDX00310A
High-Fidelity Pfu is a thermostable high fidelity DNA polymerase enzyme, isolated from Pyrococcus furiosus, with separate 10x Pfu Reaction Buffer and MgCl2. The Pfu polymerase enzyme replicates DNA in the 5´-3´ direction in the presence of magnesium, but also possesses 3´-5´ exonuclease (proofreading) activity, making it ideal for NGS library amplification.