Learn More
LAMDA BIOTECH INC Pfu DNA Polymerase
Catalog No. NC3726061
Encompass_Preferred
A high-fidelity thermostable DNA polymerase isolated from Pyrococcus furiosus, produced in E. coli through recombinant expression. This enzyme is engineered for PCR applications requiring enhanced accuracy in DNA synthesis.
Catalog No. NC3726061 Supplier LAMDA BIOTECH INC Supplier No. D151
A high-fidelity thermostable DNA polymerase isolated from Pyrococcus furiosus, produced in E. coli through recombinant expression. This enzyme is engineered for PCR applications requiring enhanced accuracy in DNA synthesis.