Learn More
Takara Bio Pfu Pyroglutamate Aminopeptidase, 10 mu
SDP
Catalog No. 50443866
Encompass
Pfu Pyroglutamate Aminopeptidase liberates the N-terminal pyroglutamic acid from proteins and peptides. This enzyme may work well with some intact, non-denatured protein. In such cases, a denaturation step may be unnecessary. This product is supplied with 5X Reaction Buffer [250 mM sodium phosphate (pH 7.0), 50 mM DTT, 5 mM EDTA]. 10mu
Catalog No. 50-443-866 Supplier Takara Bio Supplier No. 7334
Pfu Pyroglutamate Aminopeptidase liberates the N-terminal pyroglutamic acid from proteins and peptides. This enzyme may work well with some intact, non-denatured protein. In such cases, a denaturation step may be unnecessary. This product is supplied with 5X Reaction Buffer [250 mM sodium phosphate (pH 7.0), 50 mM DTT, 5 mM EDTA]. 10mu