Learn More
Agilent Technologies PFU TURBO AD 100 UNITS
Catalog No. NC9651357
Encompass
PfuTurbo DNA polymerase AD features a reformulated buffer system for increased economy with the same high performance as the original PfuTurbo DNA polymerase for robust, high-fidelity PCR.
Catalog No. NC9651357 Supplier Agilent Technologies Supplier No. 600255
PfuTurbo DNA polymerase AD features a reformulated buffer system for increased economy with the same high performance as the original PfuTurbo DNA polymerase for robust, high-fidelity PCR.