Learn More
Agilent Technologies PFU ULTRA 2-HS2X MASTER
Catalog No. NC9048637
Encompass
PFU Ultra II Hotstart PCR Master Mix is a 2X formulation of PfuUltra II Fusion HS DNA polymerase, an optimized PCR reaction buffer, magnesium, and dNTPs.
Catalog No. NC9048637 Supplier Agilent Technologies Supplier No. 600850
PFU Ultra II Hotstart PCR Master Mix is a 2X formulation of PfuUltra II Fusion HS DNA polymerase, an optimized PCR reaction buffer, magnesium, and dNTPs.