Learn More
Invivogen Human IgG1 | 20ug
SDP
Catalog No. NC1833776
Encompass
pFUSE-CHIg-hG1 is a cloning plasmid that expresses the constant region of the human IgG1 heavy chain. It contains a multiple cloning site upstream of the constant region to enable cloning of the heavy chain (CH) variable region. This plasmid is designed for antibody generation when co-transfected into mammalian cells with the light chain variable region cloned into pFUSE-CLIg. pFUSEss-CHIg-hG1 additionally features the IL2 signal sequence to allow the secretion of recombinant antibodies generated using fragment antigen-binding (Fab) or single-chain variable fragment (scFv) fragments selected from phage display libraries that lack a signal sequence.
Catalog No. NC1833776 Supplier Invivogen Supplier No. PFUSESSHCHG1
pFUSE-CHIg-hG1 is a cloning plasmid that expresses the constant region of the human IgG1 heavy chain. It contains a multiple cloning site upstream of the constant region to enable cloning of the heavy chain (CH) variable region. This plasmid is designed for antibody generation when co-transfected into mammalian cells with the light chain variable region cloned into pFUSE-CLIg. pFUSEss-CHIg-hG1 additionally features the IL2 signal sequence to allow the secretion of recombinant antibodies generated using fragment antigen-binding (Fab) or single-chain variable fragment (scFv) fragments selected from phage display libraries that lack a signal sequence.