Learn More
Agilent Technologies PFUTURBO CX DNA POLY 100 UNITS
Catalog No. NC9096856
Encompass
PfuTurbo Cx Hotstart DNA Polymerase, 100U, Specially formulated with a mutant of Pfu DNA polymerase designed to overcome uracil sensitivity for more robust performance. Stored at -20degreeC
Catalog No. NC9096856 Supplier Agilent Technologies Supplier No. 600410
PfuTurbo Cx Hotstart DNA Polymerase, 100U, Specially formulated with a mutant of Pfu DNA polymerase designed to overcome uracil sensitivity for more robust performance. Stored at -20degreeC