Learn More
Agilent Technologies PFUTURBO CX DNA POLY 500 UNITS
Catalog No. NC9746547
Encompass
Specially formulated with a mutant of Pfu DNA polymerase designed to overcome uracil sensitivity for more robust performance.
Catalog No. NC9746547 Supplier Agilent Technologies Supplier No. 600412
Specially formulated with a mutant of Pfu DNA polymerase designed to overcome uracil sensitivity for more robust performance.