Learn More
Agilent Technologies PFUTURBO DNA POLYMERASE
Catalog No. 50125949
Encompass
PfuTurbo DNA Polymerase 5 000 U (5 x 1000U) High fidelity enzyme for PCR cloning Enhances PCR product yield and length capabilities Contains: PfuTurbo DNA polymerase 10X cloned Pfu buffer
Catalog No. 50-125-949 Supplier Agilent Technologies Supplier No. 600256
Add to Cart
Add to Cart
Request a Quote
PfuTurbo DNA Polymerase 5 000 U (5 x 1000U) High fidelity enzyme for PCR cloning Enhances PCR product yield and length capabilities Contains: PfuTurbo DNA polymerase 10X cloned Pfu buffer