Learn More
Agilent Technologies PFUTURBO DNA POLYMERASEPFUTUR
Supplier:  Agilent Technologies 600252
Encompass
PfuTurbo DNA Polymerase, 500U (200 rxn), Contents: PfuTurbo DNA polymerase 10X cloned Pfu buffer, Applications: High performance PCR, For high fidelity PCR and primer-extension reactions, PCR cloning and generating blunt-end amplification p
Catalog No. 50-125-947