Learn More
Agilent Technologies PFUTURBO HOTSTART DNA POLY
Catalog No. 50125967
Encompass
PfuTurbo Hotstart DNA Polymerase 1 000 U Hot-start version of PfuTurbo DNA polymerase for added specificity Contains: PfuTurbo Hotstart DNA polymerase 10X cloned Pfu buffer Applications High-fidelity PCR cloning High-fidelity PCR
Catalog No. 50-125-967 Supplier Agilent Technologies Supplier No. 600324
PfuTurbo Hotstart DNA Polymerase 1 000 U Hot-start version of PfuTurbo DNA polymerase for added specificity Contains: PfuTurbo Hotstart DNA polymerase 10X cloned Pfu buffer Applications High-fidelity PCR cloning High-fidelity PCR