Learn More
Agilent Technologies PFUULTRA HIGH-FI DNA POLYMPFU
Supplier:  Agilent Technologies 600384
Encompass
PfuUltra high-fidelity DNA polymerase is a formulation containing a mutant of Pfu DNA polymerase that exhibits enhanced proofreading capability. Ideal for PCR cloning, site-directed mutagenesis and other applications requiring high accuracy
Catalog No. 50-125-980