Learn More
Agilent Technologies PFUULTRA HIGH-FI DNA POLYMPFU
Catalog No. 50125979
Encompass
PfuUltra high-fidelity DNA polymerase is a formulation containing a mutant of Pfu DNA polymerase that exhibits enhanced proofreading capability. Ideal for PCR cloning, site-directed mutagenesis and other applications requiring high accuracy
Catalog No. 50-125-979 Supplier Agilent Technologies Supplier No. 600382
PfuUltra high-fidelity DNA polymerase is a formulation containing a mutant of Pfu DNA polymerase that exhibits enhanced proofreading capability. Ideal for PCR cloning, site-directed mutagenesis and other applications requiring high accuracy