Learn More
Agilent Technologies PFUULTRA II HOTSTART PCR MASTE
Catalog No. NC0429470
Encompass
PfuUltra II Hotstart PCR Master Mix is a 2X formulation of PfuUltra II Fusion HS DNA polymerase, an optimized PCR reaction buffer, magnesium, and dNTPs.
Catalog No. NC0429470 Supplier Agilent Technologies Supplier No. 600852
PfuUltra II Hotstart PCR Master Mix is a 2X formulation of PfuUltra II Fusion HS DNA polymerase, an optimized PCR reaction buffer, magnesium, and dNTPs.