Learn More
Accuflow Systems Saint-Gobain PureFit Wye Insert Fitting, PVDF, 3/8 in, Barb
Catalog No. NC3363132
Encompass_Preferred
Saint-Gobain PureFit SIB Wye insert fitting in PVDF provides a chemically resistant, high-purity tubing connector for 3/8 in (nominal) tubing. The barb connection secures flexible tubing for fluid handling in bioprocessing, laboratory, and chemical delivery systems. Sold in bags of 25 for bulk installations.
  • PVDF construction for chemical resistance and high purity.
  • Wye configuration for splitting or combining fluid lines.
  • Barb connection sized for 3/8 in tubing.
  • Suitable for fluid handling, bioprocessing, and chemical delivery.
  • Sold in bags of 25 for installation efficiency.
Catalog No. NC3363132 Supplier Accuflow Systems Supplier No. PFY375PVDF
Saint-Gobain PureFit SIB Wye insert fitting in PVDF provides a chemically resistant, high-purity tubing connector for 3/8 in (nominal) tubing. The barb connection secures flexible tubing for fluid handling in bioprocessing, laboratory, and chemical delivery systems. Sold in bags of 25 for bulk installations.
  • PVDF construction for chemical resistance and high purity.
  • Wye configuration for splitting or combining fluid lines.
  • Barb connection sized for 3/8 in tubing.
  • Suitable for fluid handling, bioprocessing, and chemical delivery.
  • Sold in bags of 25 for installation efficiency.