Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Wye Connector, PVDF, 1/2 in
Catalog No. NC3363128
Encompass_Preferred
Smooth inner bore wye connector in PVDF for 1/2 in (12.7 mm) tubing. Designed to minimize dead volume and provide hygienic fluid transfer for bioprocessing and downstream applications.
  • Material: PVDF (polyvinylidene fluoride)
  • Configuration: Wye smooth inner bore connector
  • Nominal inner diameter: 1/2 in (12.7 mm)
  • Typical packaging: bag of 25 (distributor listings)
  • Tubing compatibility: compatible with Sani-Tech, C-Flex, and PharMed tubing
  • Applications: media hydration, filtration, cell culture/harvest, product sampling, downstream processing
Catalog No. NC3363128 Supplier Accuflow Systems Supplier No. PFY500PVDF
Smooth inner bore wye connector in PVDF for 1/2 in (12.7 mm) tubing. Designed to minimize dead volume and provide hygienic fluid transfer for bioprocessing and downstream applications.
  • Material: PVDF (polyvinylidene fluoride)
  • Configuration: Wye smooth inner bore connector
  • Nominal inner diameter: 1/2 in (12.7 mm)
  • Typical packaging: bag of 25 (distributor listings)
  • Tubing compatibility: compatible with Sani-Tech, C-Flex, and PharMed tubing
  • Applications: media hydration, filtration, cell culture/harvest, product sampling, downstream processing