Learn More
TARGETMOL CHEMICALS INC PG-116800 5MG
SDP
Catalog No. 502580521
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. PG-116800 (PG-530742) is a matrix metalloproteinase (MMP) inhibitor. PG-116800 can be used in studies about the treatment of osteoarthritis. purity: 98%
Catalog No. 50-258-0521 Supplier TARGETMOL CHEMICALS INC Supplier No. T283945MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. PG-116800 (PG-530742) is a matrix metalloproteinase (MMP) inhibitor. PG-116800 can be used in studies about the treatment of osteoarthritis. purity: 98%