Learn More
Medchemexpress LLC PG 34 | 1681056-62-1 | 98.0% | 547.65 | 1 ML
SDP
Catalog No. 5000360575
Encompass_Preferred
PG 34 is a weak O-GlcNAc transferase (OGT) inhibitor with an IC50 of 68 μM. It is a structural analog of OSMI-1 and is intended for research use only.
  • Weak OGT inhibitor (IC50=68 μM)
  • Structural analog of OSMI-1
  • Available in solid and solution forms
Catalog No. 50-003-60575 Supplier Medchemexpress LLC Supplier No. HY14811310MM/1ML
PG 34 is a weak O-GlcNAc transferase (OGT) inhibitor with an IC50 of 68 μM. It is a structural analog of OSMI-1 and is intended for research use only.
  • Weak OGT inhibitor (IC50=68 μM)
  • Structural analog of OSMI-1
  • Available in solid and solution forms