Learn More
Miele, Inc. PG 8583CD OIL 3AC 208V undercounter labwasher
Supplier:  Miele, Inc. 10262740
Encompass_Preferred
Designed for simplicity and sustainability, the PG 8583CD OILundercounter labwasher reduces energy consumptionand water use in the lab, while making sure mission criticalglassware is analytically clean and ready for the next set of tests. Certified with ACT Label
• High capacity throughput
(up to 128 injector nozzles)• 128 vials / 98 pipettes
• SimpleLoad modules
• Rear basket docking
• Variable Speed pump
• Auto liquid dispensing
• Multi-stage filter system
• Spray arm monitoring
• DryPlus - Hot air HEPA drying
• External DOS pump
• IO/Q Validation
Catalog No. 50-171-2997
Only null left
Add to Cart
Request a Quote