Learn More
Miele, Inc. PG 8583 OIL undercounter labwasher Base Package
Supplier:  Miele, Inc. 94004340
Encompass_Preferred
Bundle package consisting of:
1x 10254330 PG 8583 OIL
1x 12642640 A 100 Upper Basket
2x 11116960 A 302/3 Module 4x8
1x 12666330 A 150 Lower Basket
2x 11116900 A 300/3 Module 2x4
1x 10265650 ProCare Lab 11 AT
1x 10266670 ProCare Lab 30 C
1x 9195780 ProCare Universal 61
1x 11851600 CK/1 DIN 51
Catalog No. NC2738029
Add to Cart
Request a Quote