Learn More
Miele, Inc. PG 8583CD 3AC 208V undercounter labwasher
Catalog No. NC1892279
Encompass_Preferred
Designed for simplicity and sustainability, the PG 8583CD undercounter labwasher reduces energy consumptionand water use in the lab, while making sure mission criticalglassware is analytically clean and ready for the next set of tests. Certified with ACT Label
• High capacity throughput
(up to 128 injector nozzles)• 128 vials / 98 pipettes
• SimpleLoad modules
• Rear basket docking
• Variable Speed pump
• Auto liquid dispensing
• Multi-stage filter system
• Spray arm monitoring
• DryPlus - Hot air HEPA drying
• External DOS pump
• IO/Q Validation
Catalog No. NC1892279 Supplier Miele, Inc. Supplier No. 10262730
Add to Cart
Add to Cart
Request a Quote
Designed for simplicity and sustainability, the PG 8583CD undercounter labwasher reduces energy consumptionand water use in the lab, while making sure mission criticalglassware is analytically clean and ready for the next set of tests. Certified with ACT Label
• High capacity throughput
(up to 128 injector nozzles)• 128 vials / 98 pipettes
• SimpleLoad modules
• Rear basket docking
• Variable Speed pump
• Auto liquid dispensing
• Multi-stage filter system
• Spray arm monitoring
• DryPlus - Hot air HEPA drying
• External DOS pump
• IO/Q Validation