Learn More
Medchemexpress LLC PG-931 | 667430-81-1 | >99.8% | 1180.40 | 1 MG
SDP
Catalog No. 5000422298
Encompass_Preferred
PG-931 is an analog of SHU 9119 and acts as a potent agonist for the melanocortin 4 (MC4) receptor. It demonstrates higher selectivity for the hMC4R compared to the hMC3R or the hMC5R. In vivo, PG-931 can reverse hemorrhagic shock and prevent multiple organ damage.
  • Potent agonist for melanocortin 4 (MC4) receptor
  • Higher selectivity for hMC4R compared to hMC3R or hMC5R
  • Can reverse hemorrhagic shock in vivo
  • Can prevent multiple organ damage in vivo
Catalog No. 50-004-22298 Supplier Medchemexpress LLC Supplier No. HYP12081MG
PG-931 is an analog of SHU 9119 and acts as a potent agonist for the melanocortin 4 (MC4) receptor. It demonstrates higher selectivity for the hMC4R compared to the hMC3R or the hMC5R. In vivo, PG-931 can reverse hemorrhagic shock and prevent multiple organ damage.
  • Potent agonist for melanocortin 4 (MC4) receptor
  • Higher selectivity for hMC4R compared to hMC3R or hMC5R
  • Can reverse hemorrhagic shock in vivo
  • Can prevent multiple organ damage in vivo