Learn More
Medchemexpress LLC PG-931 | 667430-81-1 | 99.8% | 1180.40 | 10 MG
SDP
Catalog No. 5000422331
Encompass_Preferred
PG-931 is an analog of SHU 9119 and a potent, selective melanocortin 4 (MC4) receptor agonist. This compound has demonstrated the ability to reverse hemorrhagic shock and prevent multiple organ damage in vivo.
  • Potent melanocortin 4 (MC4) receptor agonist with an IC50 of 0.58 nM.
  • More selective for MC4R than for hMC3R (IC50=55 nM) or hMC5R (IC50=2.4 nM).
  • Can reverse hemorrhagic shock and prevent multiple organ damage in vivo.
  • Exhibits anti-shock effects at nanomolar doses.
Catalog No. 50-004-22331 Supplier Medchemexpress LLC Supplier No. HYP120810MG
PG-931 is an analog of SHU 9119 and a potent, selective melanocortin 4 (MC4) receptor agonist. This compound has demonstrated the ability to reverse hemorrhagic shock and prevent multiple organ damage in vivo.
  • Potent melanocortin 4 (MC4) receptor agonist with an IC50 of 0.58 nM.
  • More selective for MC4R than for hMC3R (IC50=55 nM) or hMC5R (IC50=2.4 nM).
  • Can reverse hemorrhagic shock and prevent multiple organ damage in vivo.
  • Exhibits anti-shock effects at nanomolar doses.