Learn More
Medchemexpress LLC HY-103369 1mg Medchemexpress, PG01 CAS:853138-65-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1033691MG
Encompass_Preferred
Medchemexpress, HY-103369 1mg PG01 CAS:853138-65-5 PG01 is a potent CFTR Cl- channel potentiator. PG01 can correct gating defects of CFTR mutants, is effective on b>E193K, G970R and G551D (CFTR mutants) with Kd values of 0.22 μM, 0.45 μM and 1.94 μM, respectively. PG01 is also effective on ΔF508 (Ka of 0.3 μM). PG01 increases ΔF508-CFTR Cl- current after adding Forskolin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8089