Learn More
MSC Intertape Masking Tape, 2 inch, 60 yd, 24 Rolls
Catalog No. 501305403
Encompass
Intertape masking tape is designed for a variety of surfaces including glass, urethane-finished, lacquer-coated, and unfinished wood. It features a tear-resistant backing and paint flake-resistant properties, suitable for masking during painting and other applications.
  • For glass, urethane-finished, lacquer-coated, and unfinished wood surfaces
  • Tear-resistant backing
  • Paint flake-resistant
  • Conformable
  • Lacquer-resistant
  • Recommended for hot to warm removal, can be removed cold
  • Removes cleanly after exposure to temperatures up to 325 °F for 30 min
Catalog No. 50-130-5403 Supplier MSC Supplier No. 65167769
Intertape masking tape is designed for a variety of surfaces including glass, urethane-finished, lacquer-coated, and unfinished wood. It features a tear-resistant backing and paint flake-resistant properties, suitable for masking during painting and other applications.
  • For glass, urethane-finished, lacquer-coated, and unfinished wood surfaces
  • Tear-resistant backing
  • Paint flake-resistant
  • Conformable
  • Lacquer-resistant
  • Recommended for hot to warm removal, can be removed cold
  • Removes cleanly after exposure to temperatures up to 325 °F for 30 min