Learn More
BEZWADA BIOMEDICAL LLC Poly(glycolide-co-caprolactone), Glycolide:caprolactone 55:45, viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Catalog No. 501904484
Encompass_Preferred
Copolymers of glycolide with caprolactone have been developed to reduce the stiffness and increase the absorption time of the polymer. These copolymers have found numerous applications in medical device and controlled drug delivery applications.
Catalog No. 50-190-4484 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 2154
Copolymers of glycolide with caprolactone have been developed to reduce the stiffness and increase the absorption time of the polymer. These copolymers have found numerous applications in medical device and controlled drug delivery applications.