Learn More
BEZWADA BIOMEDICAL LLC Poly(glycolide-co-caprolactone), Glycolide:caprolactone 60:40, viscosity 1.2 to 2.0 dl/g, 5.0 GM
SDP
Catalog No. 501904483
Encompass_Preferred
Copolymers of glycolide with caprolactone have been developed to reduce the stiffness and increase the absorption time of the polymer. These copolymers have found numerous applications in medical device and controlled drug delivery applications.
Catalog No. 50-190-4483 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 2164
Copolymers of glycolide with caprolactone have been developed to reduce the stiffness and increase the absorption time of the polymer. These copolymers have found numerous applications in medical device and controlled drug delivery applications.