Learn More
BEZWADA BIOMEDICAL LLC Poly(glycolide-co-caprolactone), Glycolide:caprolactone 75:25, viscosity 1.4 to 2.0 dl/g, 5.0 GM
SDP
Catalog No. 501904482
Encompass_Preferred
Copolymers of glycolide with caprolactone have been developed to reduce the stiffness and increase the absorption time of the polymer. These copolymers have found numerous applications in medical device and controlled drug delivery applications.
Catalog No. 50-190-4482 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 2172
Copolymers of glycolide with caprolactone have been developed to reduce the stiffness and increase the absorption time of the polymer. These copolymers have found numerous applications in medical device and controlled drug delivery applications.