Learn More
BEZWADA BIOMEDICAL LLC Poly(glycolide-co-L-lactide) Glycolide:L-Lactide 95:5, viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Catalog No. 501904479
Encompass_Preferred
In order to expand the utility of lactide and glycolide monomers beyond its homopolymer use and to extend the bandwith of homopolymer properties, a number of copolymers of glycolide with both L(-)-lactide and DL-lactide have been developed. These copolymers have found numerous applications in medical device and controlled drug delivery applications. For example, a copolymer of 90% glycolide and 10% l-lactide was developed by Ethicon as an absorbable suture material under the trade name VicrylTM. Similarly, MonocrylTM sold as a monofilament suture by Ethicon, Inc. is a block copolymer of -caprolactone with glycolide, and has reduced stiffness compared to DexonTM which is a prepared from polyglycolide homopolymer.Different copolymers of glycolide can be prepared for varying physical properties and absorption. The polymers are used in medical devices such as sutures, screws, staples and clips.
Catalog No. 50-190-4479 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 2395
In order to expand the utility of lactide and glycolide monomers beyond its homopolymer use and to extend the bandwith of homopolymer properties, a number of copolymers of glycolide with both L(-)-lactide and DL-lactide have been developed. These copolymers have found numerous applications in medical device and controlled drug delivery applications. For example, a copolymer of 90% glycolide and 10% l-lactide was developed by Ethicon as an absorbable suture material under the trade name VicrylTM. Similarly, MonocrylTM sold as a monofilament suture by Ethicon, Inc. is a block copolymer of -caprolactone with glycolide, and has reduced stiffness compared to DexonTM which is a prepared from polyglycolide homopolymer.Different copolymers of glycolide can be prepared for varying physical properties and absorption. The polymers are used in medical devices such as sutures, screws, staples and clips.