Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PGAM1-IN-1 | 2438637-65-9 | 99.8% | 10 MG
SDP

Catalog No. 5000339420
Encompass_Preferred

PGAM1-IN-1 is a phosphoglycerate mutase 1 (PGAM1) inhibitor with an IC50 of 6.4 μM. It inhibits H1299 proliferation with an IC50 of 14.1±1.9 μM. This product is for research use only and not sold to patients.

  • Inhibits phosphoglycerate mutase 1 (PGAM1) with an IC50 of 6.4 μM.
  • Inhibits H1299 proliferation with an IC50 of 14.1±1.9 μM.
  • Molecular weight: 435.81.
  • Chemical formula: C19H11ClFNO6S.
  • Appearance: solid.
  • Color: light yellow to yellow.
  • For research use only.

Catalog No. 50-003-39420 Supplier Medchemexpress LLC Supplier No. HY12868110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PGAM1-IN-1 is a phosphoglycerate mutase 1 (PGAM1) inhibitor with an IC50 of 6.4 μM. It inhibits H1299 proliferation with an IC50 of 14.1±1.9 μM. This product is for research use only and not sold to patients.

  • Inhibits phosphoglycerate mutase 1 (PGAM1) with an IC50 of 6.4 μM.
  • Inhibits H1299 proliferation with an IC50 of 14.1±1.9 μM.
  • Molecular weight: 435.81.
  • Chemical formula: C19H11ClFNO6S.
  • Appearance: solid.
  • Color: light yellow to yellow.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.