Learn More
Medchemexpress LLC PGAM1-IN-2 | 2438637-66-0 | 98.0% | 10 MG
SDP
Catalog No. 5000338287
Encompass_Preferred
PGAM1-IN-2 is a phosphoglycerate mutase 1 (PGAM1) inhibitor with an IC50 of 2.1 μM. This compound inhibits H1299 proliferation with an IC50 of 33.8±6.1 μM.
  • Potent phosphoglycerate mutase 1 inhibitor
  • Inhibits H1299 proliferation
  • Suitable for research applications
Catalog No. 50-003-38287 Supplier Medchemexpress LLC Supplier No. HY12868210MG
PGAM1-IN-2 is a phosphoglycerate mutase 1 (PGAM1) inhibitor with an IC50 of 2.1 μM. This compound inhibits H1299 proliferation with an IC50 of 33.8±6.1 μM.
  • Potent phosphoglycerate mutase 1 inhibitor
  • Inhibits H1299 proliferation
  • Suitable for research applications