Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pgam1-in-2 | 2438637-66-0 | 98.0% | 100 MG
SDP

Catalog No. 5000340020
Encompass_Preferred

PGAM1-IN-2 is an inhibitor of phosphoglycerate mutase 1 (PGAM1) with an IC50 of 2.1 μM. It is for research use only and inhibits H1299 proliferation with an IC50 of 33.8±6.1 μM.

  • Inhibits phosphoglycerate mutase 1 (PGAM1) with an IC50 of 2.1 μM.
  • Inhibits H1299 proliferation with an IC50 of 33.8±6.1 μM.
  • Provided as a solid.
  • For research use only.
  • Target: Phosphatase.
  • Pathway: Metabolic enzyme/protease.
  • Storage for powder: -20°C for 3 years, 4°C for 2 years.
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-003-40020 Supplier Medchemexpress LLC Supplier No. HY128682100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PGAM1-IN-2 is an inhibitor of phosphoglycerate mutase 1 (PGAM1) with an IC50 of 2.1 μM. It is for research use only and inhibits H1299 proliferation with an IC50 of 33.8±6.1 μM.

  • Inhibits phosphoglycerate mutase 1 (PGAM1) with an IC50 of 2.1 μM.
  • Inhibits H1299 proliferation with an IC50 of 33.8±6.1 μM.
  • Provided as a solid.
  • For research use only.
  • Target: Phosphatase.
  • Pathway: Metabolic enzyme/protease.
  • Storage for powder: -20°C for 3 years, 4°C for 2 years.
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.