Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci PGAM5 (CT) Peptide
SDP

Catalog No. 16595757
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-757 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-757 Supplier ProSci Supplier No. 9517P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

PGAM5 (CT) Peptideis used for blocking the activity of thePGAM5 (CT) Antibody

PGAM5 Antibody: PGAM5 is a central mediator for programmed necrosis induced by TNF and may be a regulator of mitochondrial dynamics. It displays phosphatase activity for serine/threonine residues, dephosphorylates and activates MAP3K5 kinase. It has apparently no phosphoglycerate mutase activity and is a substrate for a KEAP1-dependent ubiquitin ligase complex. It is also contributes to the repression of NFE2L2-dependent gene expression

Specifications

Gene ID (Entrez) 192111
Species Human
Accession Number Q96HS1
Common Name PGAM family member 5, mitochondrial serine/threonine protein phosphatase
Concentration 200 μg/mL
Content And Storage Store PGAM5 (CT) Peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol PGAM5
Quantity 0.05 mg
Sequence 14 amino acids near the carboxy terminus of PGAM5 (CT).
Synonym Serine/threonine-protein phosphatase PGAM5 mitochondrial, Bcl-XL-binding protein v68, Phosphoglycerate mutase family member 5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.