Learn More
Medchemexpress LLC PGAM5 protein, human (His) | >95.0% | 50UG
SDP
Catalog No. 5000174311
Encompass_Preferred
Recombinant human PGAM5 protein with a C-terminal His tag, expressed in E. coli and supplied lyophilized. PGAM5 is a mitochondrial serine/threonine phosphatase involved in regulating mitochondrial dynamics, mitophagy, necroptosis, and antioxidant responses. Supplied as 50 μg of material with storage and stability information for biochemical and cell biology applications.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Expressed in E. coli for recombinant protein production.
  • C-terminal His tag enables affinity purification and detection.
  • Supplied as 50 μg lyophilized material for flexible assay use.
  • Formulated with trehalose for stability during lyophilization.
  • Storage recommended at -20°C; reconstituted stability short term at 4°C.
Catalog No. 50-001-74311 Supplier Medchemexpress LLC Supplier No. HYP70286950UG
Recombinant human PGAM5 protein with a C-terminal His tag, expressed in E. coli and supplied lyophilized. PGAM5 is a mitochondrial serine/threonine phosphatase involved in regulating mitochondrial dynamics, mitophagy, necroptosis, and antioxidant responses. Supplied as 50 μg of material with storage and stability information for biochemical and cell biology applications.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Expressed in E. coli for recombinant protein production.
  • C-terminal His tag enables affinity purification and detection.
  • Supplied as 50 μg lyophilized material for flexible assay use.
  • Formulated with trehalose for stability during lyophilization.
  • Storage recommended at -20°C; reconstituted stability short term at 4°C.