Learn More
Aviva Systems Biology Corp PGBD1 Antibody - C-terminal region (ARP39704_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39704_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PGBD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PGBD1. Alternate names: SCAND4, HUCEP-4, dJ874C20.4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-57144