Learn More
Aviva Systems Biology Corp PGBD2 Antibody - middle region (ARP54446_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54446_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PGBD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PGBD2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rabbit
Catalog No. 50-014-61208