Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PGC1 alpha+beta Antibody (YA3458, Rabbit) | 50 UL
SDP

Catalog No. 5000443691
Encompass_Preferred

PGC1 alpha+beta Antibody (YA3458) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to PGC1 alpha+beta.

  • Antibody
  • Regulatory status: RUO
  • Host: Rb
  • Genebank: 10891 (Gene ID) (Accession)
  • Isotype: IgG
  • Format: Not specified
  • Storage conditions: Stored at -20°C for 1 year
  • Shipping details: Shipping with blue ice

Catalog No. 50-004-43691 Supplier Medchemexpress LLC Supplier No. HYP8372850UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PGC1 alpha+beta Antibody (YA3458) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to PGC1 alpha+beta.

  • Antibody
  • Regulatory status: RUO
  • Host: Rb
  • Genebank: 10891 (Gene ID) (Accession)
  • Isotype: IgG
  • Format: Not specified
  • Storage conditions: Stored at -20°C for 1 year
  • Shipping details: Shipping with blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.