Learn More
TARGETMOL CHEMICALS INC PGD2-IN-1 5MG
SDP
Catalog No. 502579860
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PGD2-IN-1 (PGD2-inhibitor) is an effective DP receptor antagonist (IC500.3 nM) a compound that can inhibit prostaglandin D2 (PGD2) signaling. PGD2-IN-1 specifically targets and regulates the activity of PGD2 receptors or enzymes involved IN PGD2 synthesis or metabolism. purity: 98%
Catalog No. 50-257-9860 Supplier TARGETMOL CHEMICALS INC Supplier No. T4624
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PGD2-IN-1 (PGD2-inhibitor) is an effective DP receptor antagonist (IC500.3 nM) a compound that can inhibit prostaglandin D2 (PGD2) signaling. PGD2-IN-1 specifically targets and regulates the activity of PGD2 receptors or enzymes involved IN PGD2 synthesis or metabolism. purity: 98%