Learn More
Medchemexpress LLC HY-101430 5mg Medchemexpress, PGD2-IN-1 CAS:885066-67-1 Purity:>98%
SDP
Catalog No. 501932127
Encompass_Preferred
Medchemexpress, HY-101430 5mg PGD2-IN-1 CAS:885066-67-1 PGD2-IN-1 is an antagonist of DP extracted from patent WO 2006044732 A2, example 15 (d); has an IC 50 of 0.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2127 Supplier Medchemexpress LLC Supplier No. HY1014305MG
Medchemexpress, HY-101430 5mg PGD2-IN-1 CAS:885066-67-1 PGD2-IN-1 is an antagonist of DP extracted from patent WO 2006044732 A2, example 15 (d); has an IC 50 of 0.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.