Learn More
TARGETMOL CHEMICALS INC PGD2-IN-1 5mg
SDP
Catalog No. NC1634658
Encompass_Preferred
PGD2-IN-1 is an effective DP receptor antagonist (IC50:0.3 nM), a compound that can inhibit prostaglandin D2 (PGD2) signaling. PGD2-IN-1 specifically targets and regulates the activity of PGD2 receptors or enzymes involved IN PGD2 synthesis or metabolism.
Catalog No. NC1634658 Supplier TARGETMOL CHEMICALS INC Supplier No. T46245MG
Add to Cart
Add to Cart
PGD2-IN-1 is an effective DP receptor antagonist (IC50:0.3 nM), a compound that can inhibit prostaglandin D2 (PGD2) signaling. PGD2-IN-1 specifically targets and regulates the activity of PGD2 receptors or enzymes involved IN PGD2 synthesis or metabolism.