Learn More
Enzo Life Sciences PGE1 ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900005
Encompass_Preferred
The PGE1 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. Assay up to 38 samples in duplicate per kit with excellent detection sensitivity. Alternative name: Prostaglandin E1. Assay Time: <3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .
Catalog No. 50-200-6826